| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023033.hg.1 |
ENST00000257414.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023033.hg.1 |
ENST00000282394.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023033.hg.1 |
ENST00000343348.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023033.hg.1 |
ENST00000510263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023033.hg.1 |
NM_020796.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023033.hg.1 |
TC05001700.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023032.hg.1 |
ENST00000257414.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023032.hg.1 |
ENST00000282394.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023032.hg.1 |
ENST00000343348.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023032.hg.1 |
ENST00000510263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023032.hg.1 |
NM_020796.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023032.hg.1 |
TC05001700.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000257414.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000343348.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000506114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000510263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000515129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
NM_001300780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
NM_020796.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000257414.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000343348.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000506114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000510263.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
ENST00000515129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
NM_001300780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
NM_020796.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181432.hg.1 |
TC0500011775.hg.1 |
Clariom D TC |