Assay Details
| Gene Symbol: SEMA6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005272042.3 | XP_005272099.2 | 15 - 16 | 1993 | 115 | |
| GenBank mRNA | AK027439.1 | - | 3 - 4 | 371 | 115 | |
More Information
Set Membership: |
|

SEMA6A
Genomic Map