Assay Details
| Gene Symbol: SEMA6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005272042.3 | XP_005272099.2 | 16 - 17 | 2059 | 102 | |
| GenBank mRNA | AK027439.1 | - | 4 - 5 | 437 | 102 | |
More Information
Set Membership: |
|

SEMA6A
Genomic Map