Assay Details
| Gene Symbol: SEMA6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300780.1 | NP_001287709.1 | 18 - 19 | 2561 | 95 | |
| GenBank mRNA | AB037789.1 | - | 18 - 19 | 2030 | 95 | |
More Information
Set Membership: |
|

SEMA6A
Genomic Map