Assay Details
| Gene Symbol: IMPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282368.1 | NP_001269297.1 | 8 - 9 | 847 | 125 | |
| NM_001563.3 | NP_001554.2 | 9 - 10 | 1081 | 125 | ||
| GenBank mRNA | AF047492.2 | - | 9 - 10 | 1021 | 125 | |
| BC117450.1 | - | 9 - 10 | 911 | 40126001 | 125 | |
| BC117452.1 | - | 9 - 10 | 911 | 40126003 | 125 | |
| CR749572.1 | - | 9 - 10 | 3323 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

IMPG1
Genomic Map