Assay Details
| Gene Symbol: IMPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282368.1 | NP_001269297.1 | 2 - 3 | 426 | 132 | |
| NM_001563.3 | NP_001554.2 | 3 - 4 | 660 | 132 | ||
| GenBank mRNA | AF047492.2 | - | 3 - 4 | 600 | 132 | |
| BC117450.1 | - | 3 - 4 | 490 | 40126001 | 132 | |
| BC117452.1 | - | 3 - 4 | 490 | 40126003 | 132 | |
| CA391789.1 | - | 2 - 3 | 396 | 132 | ||
| CR749572.1 | - | 2 - 3 | 426 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001889.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022983.hg.1 | ENST00000369950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022983.hg.1 | ENST00000369963.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022983.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022983.hg.1 | TC06001889.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022982.hg.1 | ENST00000369950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022982.hg.1 | ENST00000369963.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022982.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022982.hg.1 | TC06001889.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185059.hg.1 | TC0600012329.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369952.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000369963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | ENST00000611179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001282368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012329.hg.1 | NM_001563.3 | Refseq |

IMPG1
Genomic Map