Assay Details
| Gene Symbol: TAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017002188.1 | XP_016857677.1 | 1 - 2 | 278 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017002188.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
locomotory behavior
spinal cord association neuron differentiation
hemopoiesis
erythrocyte differentiation
megakaryocyte differentiation
platelet formation
basophil differentiation
positive regulation of protein complex assembly
embryonic hemopoiesis
megakaryocyte development
regulation of cell proliferation
erythrocyte maturation
cell fate commitment
positive regulation of erythrocyte differentiation
positive regulation of chromatin assembly or disassembly
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of cell division
astrocyte fate commitment
definitive hemopoiesis
hemangioblast cell differentiation
hematopoietic stem cell differentiation
regulation of mast cell differentiation
regulation of stem cell population maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
enzyme binding
histone deacetylase binding
transcription regulatory region DNA binding
protein heterodimerization activity
E-box binding
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
locomotory behavior
spinal cord association neuron differentiation
hemopoiesis
erythrocyte differentiation
megakaryocyte differentiation
platelet formation
basophil differentiation
positive regulation of protein complex assembly
embryonic hemopoiesis
megakaryocyte development
regulation of cell proliferation
erythrocyte maturation
cell fate commitment
positive regulation of erythrocyte differentiation
positive regulation of chromatin assembly or disassembly
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of cell division
astrocyte fate commitment
definitive hemopoiesis
hemangioblast cell differentiation
hematopoietic stem cell differentiation
regulation of mast cell differentiation
regulation of stem cell population maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
enzyme binding
histone deacetylase binding
transcription regulatory region DNA binding
protein heterodimerization activity
E-box binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000464796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000464796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000465912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000465912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000465912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000481091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000481091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | ENST00000481091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000294339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000371884.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000459729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000464796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000464796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000465912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000465912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000465912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000481091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000481091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | ENST00000481091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001287347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_001290406.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018439.hg.1 | NM_003189.5 | Refseq |

TAL1
Genomic Map