Assay Details
| Gene Symbol: DNAJC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013238.2 | NP_037370.2 | 2 - 3 | 563 | 157 | |
| GenBank mRNA | AF126743.1 | - | 3 - 4 | 585 | 157 | |
| AK311866.1 | - | 2 - 3 | 199 | 157 | ||
| AY248898.1 | - | 2 - 3 | 563 | 157 | ||
| AY512565.1 | - | 2 - 3 | 256 | 157 | ||
| BC095400.1 | - | 2 - 3 | 182 | 30530999 | 157 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000157.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13001836.hg.1 | ENST00000379221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001836.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001836.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001836.hg.1 | TC13000157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001834.hg.1 | ENST00000379221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001834.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001834.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001834.hg.1 | TC13000157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001832.hg.1 | ENST00000379221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001832.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001832.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001832.hg.1 | TC13000157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011529.hg.1 | ENST00000416693.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001833.hg.1 | ENST00000379221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001833.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001833.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001833.hg.1 | TC13000157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001835.hg.1 | ENST00000379221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001835.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001835.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001835.hg.1 | TC13000157.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300166462.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166462.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166462.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166462.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166462.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166462.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166462.hg.1 | TC1300009954.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011529.hg.1 | ENST00000416693.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009954.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166465.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166465.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166465.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166465.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166465.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166465.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166465.hg.1 | TC1300009954.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300166464.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166464.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166464.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166464.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166464.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166464.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166464.hg.1 | TC1300009954.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300166466.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166466.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166466.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166466.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166466.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166466.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166466.hg.1 | TC1300009954.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300166461.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166461.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166461.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166461.hg.1 | ENST00000379221.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166461.hg.1 | ENST00000474320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166461.hg.1 | NM_013238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166461.hg.1 | TC1300009954.hg.1 | Clariom D TC |

DNAJC15
Genomic Map