| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
230488_PM_s_at |
- |
- |
| Affymetrix Human Clariom D Assay |
PSR0900181200.hg.1 |
ENST00000425189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181200.hg.1 |
NR_102735.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181200.hg.1 |
ENST00000425189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181200.hg.1 |
NR_102735.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181200.hg.1 |
TC0900011859.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900181203.hg.1 |
ENST00000425189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181203.hg.1 |
NR_102735.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181203.hg.1 |
ENST00000425189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181203.hg.1 |
NR_102735.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181203.hg.1 |
TC0900011859.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900181204.hg.1 |
NR_102735.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181204.hg.1 |
NR_102735.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181204.hg.1 |
TC0900011859.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
230488_PM_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
230488_s_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230488_s_at2 |
- |
- |
| Human Genome U95 Set |
87446_PM_at |
- |
- |
| Human Genome U95 Set |
87446_at |
- |
- |
| Human Genome U133 Set |
230488_PM_s_at |
- |
- |
| Human Genome U133 Set |
230488_s_at |
- |
- |