Assay Details
| Gene Symbol: MATN1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128076.1 | - | 2 | 1475 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000414763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000443076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000443076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414532.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000414763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000443076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000443076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | ENST00000454613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | NR_034182.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007614.hg.1 | NR_034182.1 | Refseq |

MATN1-AS1
Genomic Map