Assay Details
| Gene Symbol: ZIM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110744.1 | 1 - 2 | 89 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 835 | ZNF835 | BC37295_3 | -93.5kb | Anti-sense | Chr. 19 - 56662652 - 56671783 on NCBI Build GRCh38 |
| Downstream | paternally expressed 3 | PEG3 |
PW1, ZKSCAN22, ZNF904, ZSCAN24 |
20.0kb | Anti-sense | Chr. 19 - 56810077 - 56840726 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ZIM2-AS1
Genomic Map