Assay Details
| Gene Symbol: COL4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK000715.1 | - | 4 - 5 | 437 | 111 | |
| X05562.1 | - | 25 - 26 | 2268 | 111 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000463084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000463084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000463084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000494852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000494852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000494852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000617564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000617564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |

COL4A2
Genomic Map