Assay Details
| Gene Symbol: COL4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK000715.1 | - | 5 - 6 | 493 | 85 | |
| X05562.1 | - | 26 - 27 | 2324 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COL4A2| Gene Symbol: COL4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK000715.1 | - | 5 - 6 | 493 | 85 | |
| X05562.1 | - | 26 - 27 | 2324 | 85 | ||
Set Membership: |
|