Assay Details
| Gene Symbol: COL4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025912.1 | - | 4 - 5 | 448 | 126 | |
| AK294920.1 | - | 8 - 9 | 775 | 126 | ||
| BX648393.1 | - | 5 - 6 | 514 | 126 | ||
| BY795218.2 | - | 5 - 6 | 589 | 126 | ||
| M24766.1 | - | 3 - 4 | 226 | 126 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000360467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000400163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000462309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000463084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000463084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000463084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000478681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000480771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000483683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000494852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000494852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000494852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000617564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | ENST00000617564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008010.hg.1 | NM_001846.3 | Refseq |

COL4A2
Genomic Map