Assay Details
| Gene Symbol: COL4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025912.1 | - | 7 - 8 | 734 | 54 | |
| AK294920.1 | - | 11 - 12 | 1061 | 54 | ||
| BX648393.1 | - | 8 - 9 | 800 | 54 | ||
| M24766.1 | - | 6 - 7 | 512 | 54 | ||
More Information
Set Membership: |
|

COL4A2
Genomic Map