Assay Details
| Gene Symbol: SMYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264156.2 | XP_005264213.1 | 4 - 5 | 699 | 86 | |
| GenBank mRNA | AY518934.1 | - | 1 - 2 | 49 | 86 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
heart development
positive regulation of myotube differentiation
histone lysine methylation
skeletal muscle cell differentiation
positive regulation of myoblast differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription corepressor activity
protein binding
histone-lysine N-methyltransferase activity
metal ion binding
chromatin remodeling
transcription, DNA-templated
heart development
positive regulation of myotube differentiation
histone lysine methylation
skeletal muscle cell differentiation
positive regulation of myoblast differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription corepressor activity
protein binding
histone-lysine N-methyltransferase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000438570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000438570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000438570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000468008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000468008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | ENST00000468008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000419482.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000438570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000438570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000438570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000444564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000468008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000468008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | ENST00000468008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008334.hg.1 | NM_198274.3 | Refseq |

SMYD1
Genomic Map