| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004145.hg.1 |
ENST00000187762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004145.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004145.hg.1 |
TC19000323.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900152808.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152808.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900152808.hg.1 |
TC1900007290.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900152808.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152808.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000187762.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000595452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
ENST00000599479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007290.hg.1 |
NM_024074.2 |
Refseq |