Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012775.1 | XP_016868264.1 | 2 - 3 | 355 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012775.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000307471.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000409765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000410069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000448353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000472407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000484553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000485233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | ENST00000486109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006928.hg.1 | NM_138771.3 | Refseq |

CCDC126
Genomic Map