Assay Details
| Gene Symbol: DHX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001358.2 | NP_001349.2 | 8 - 9 | 1648 | 105 | |
| GenBank mRNA | AB001636.1 | - | 8 - 9 | 1641 | 105 | |
| AF279891.1 | - | 8 - 9 | 1480 | 105 | ||
| AK303506.1 | - | 8 - 9 | 1588 | 105 | ||
| BC035974.1 | - | 8 - 9 | 1635 | 4806964 | 105 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001086.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04014935.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014935.hg.1 | ENST00000535946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014935.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014935.hg.1 | TC04001086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000476316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014936.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014936.hg.1 | ENST00000535946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014936.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014936.hg.1 | TC04001086.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400170153.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170153.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170153.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400170153.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170153.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170153.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400170153.hg.1 | TC0400010251.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000476316.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400170154.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170154.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170154.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400170154.hg.1 | ENST00000336812.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170154.hg.1 | ENST00000503562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400170154.hg.1 | NM_001358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400170154.hg.1 | TC0400010251.hg.1 | Clariom D TC |

DHX15
Genomic Map