| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023094.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023094.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023094.hg.1 |
TC07001535.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023091.hg.1 |
ENST00000005180.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023091.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023091.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023091.hg.1 |
TC07001535.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023090.hg.1 |
ENST00000005180.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023090.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023090.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023090.hg.1 |
TC07001535.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023092.hg.1 |
ENST00000005180.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023092.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023092.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023092.hg.1 |
TC07001535.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023089.hg.1 |
ENST00000005180.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023089.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023089.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023089.hg.1 |
TC07001535.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700182279.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182279.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182279.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182279.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182279.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182279.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182279.hg.1 |
TC0700011560.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700182280.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182280.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182280.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182280.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182280.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182280.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182280.hg.1 |
TC0700011560.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700182281.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182281.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182281.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182281.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182281.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182281.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182281.hg.1 |
TC0700011560.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700182283.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182283.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182283.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182283.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182283.hg.1 |
TC0700011560.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700182277.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182277.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182277.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182277.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182277.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182277.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182277.hg.1 |
TC0700011560.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700182278.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182278.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182278.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182278.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182278.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182278.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182278.hg.1 |
TC0700011560.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000005180.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
ENST00000394905.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011560.hg.1 |
NM_006072.4 |
Refseq |