Assay Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181714.3 | NP_859065.2 | 1 - 2 | 310 | 156 | |
| GenBank mRNA | BC050327.1 | - | 1 - 2 | 310 | 4823983 | 156 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001899.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC050327.1,NM_181714.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06023079.hg.1 | ENST00000392959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023079.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023079.hg.1 | TC06001899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023081.hg.1 | ENST00000369846.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023081.hg.1 | ENST00000392959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023081.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023081.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023081.hg.1 | TC06001899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023080.hg.1 | ENST00000392959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023080.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023080.hg.1 | TC06001899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023082.hg.1 | ENST00000369846.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023082.hg.1 | ENST00000392959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023082.hg.1 | ENST00000467898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023082.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023082.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023082.hg.1 | TC06001899.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185287.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185287.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185287.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185287.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185287.hg.1 | TC0600012372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600185288.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185288.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185288.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185288.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185288.hg.1 | TC0600012372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185289.hg.1 | TC0600012372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185290.hg.1 | TC0600012372.hg.1 | Clariom D TC |

LCA5
Genomic Map