Assay Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122769.2 | NP_001116241.1 | 2 - 3 | 637 | 102 | |
| NM_181714.3 | NP_859065.2 | 3 - 4 | 792 | 102 | ||
| XM_005248665.4 | XP_005248722.1 | 2 - 3 | 651 | 102 | ||
| XM_011535504.1 | XP_011533806.1 | 3 - 4 | 704 | 102 | ||
| GenBank mRNA | AK299314.1 | - | 2 - 3 | 608 | 102 | |
| BC050327.1 | - | 3 - 4 | 792 | 4823983 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001899.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06023077.hg.1 | ENST00000369846.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023077.hg.1 | ENST00000392959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023077.hg.1 | ENST00000467898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023077.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023077.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023077.hg.1 | TC06001899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185284.hg.1 | TC0600012372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012372.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | ENST00000369846.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | ENST00000392959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | ENST00000467898.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | NM_001122769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | NM_181714.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600185285.hg.1 | TC0600012372.hg.1 | Clariom D TC |

LCA5
Genomic Map