Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC050051.1 | - | 1 - 2 | 123 | 5278133 | 130 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050051.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000279396.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000561899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000566848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569445.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000569481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000570255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000575829.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | ENST00000602948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_001083613.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011380.hg.1 | NM_194280.3 | Refseq |

TMEM219
Genomic Map