Assay Details
| Gene Symbol: UTP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014503.2 | NP_055318.2 | 40 - 41 | 5262 | 101 | |
| GenBank mRNA | AJ006778.1 | - | 40 - 41 | 5250 | 101 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000781.hg.1
Set Membership: |
|

UTP20
Genomic Map