Assay Details
| Gene Symbol: UTP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014503.2 | NP_055318.2 | 58 - 59 | 7817 | 86 | |
| GenBank mRNA | AF072718.1 | - | 16 - 17 | 2016 | 86 | |
| AJ006778.1 | - | 58 - 59 | 7805 | 86 | ||
| AK000333.1 | - | 13 - 14 | 1600 | 86 | ||
| AK297739.1 | - | 9 - 10 | 1018 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000781.hg.1
Set Membership: |
|

UTP20
Genomic Map