Assay Details
| Gene Symbol: XAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020196.2 | NP_064581.2 | 15 - 16 | 2119 | 73 | |
| GenBank mRNA | AB026111.1 | - | 15 - 16 | 2108 | 73 | |
| AB033003.1 | - | 13 - 14 | 1795 | 73 | ||
| AF226051.1 | - | 15 - 16 | 2122 | 73 | ||
| AF258567.1 | - | 15 - 16 | 2114 | 73 | ||
| AF272147.1 | - | 15 - 16 | 2129 | 73 | ||
| AK025858.1 | - | 10 - 11 | 1402 | 73 | ||
| BC007208.1 | - | 15 - 16 | 2101 | 2823235 | 73 | |
| BC008778.1 | - | 10 - 11 | 1434 | 73 | ||
| CR749864.1 | - | 12 - 13 | 1648 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001114.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19015300.hg.1 | ENST00000358368.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015300.hg.1 | ENST00000534844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015300.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015300.hg.1 | TC19001114.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000596134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000596134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000596134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015301.hg.1 | ENST00000358368.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015301.hg.1 | ENST00000534844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015301.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015301.hg.1 | TC19001114.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | TC1900009487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176248.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000596134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000596134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000596134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009487.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | NM_020196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | TC1900009487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | ENST00000358368.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | ENST00000595288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | ENST00000600230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176250.hg.1 | NM_020196.2 | Refseq |

XAB2
Genomic Map