Assay Details
| Gene Symbol: ELAVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001419.2 | NP_001410.2 | 3 - 4 | 448 | 72 | |
| GenBank mRNA | AK301013.1 | - | 3 - 4 | 450 | 72 | |
| BC003376.2 | - | 3 - 4 | 411 | 2901220 | 72 | |
| BT009793.1 | - | 2 - 3 | 281 | 72 | ||
| BU542524.1 | - | 3 - 4 | 449 | 72 | ||
| U38175.1 | - | 3 - 4 | 399 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001125.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
multicellular organism development
regulation of mRNA stability
positive regulation of translation
mRNA stabilization
negative regulation of gene silencing by miRNA
3'-UTR-mediated mRNA stabilization
regulation of stem cell population maintenance
nucleotide binding
RNA binding
double-stranded RNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
AU-rich element binding
protein kinase binding
mRNA 3'-UTR AU-rich region binding
protein homodimerization activity
poly(A) RNA binding
mRNA splicing, via spliceosome
multicellular organism development
regulation of mRNA stability
positive regulation of translation
mRNA stabilization
negative regulation of gene silencing by miRNA
3'-UTR-mediated mRNA stabilization
regulation of stem cell population maintenance
nucleotide binding
RNA binding
double-stranded RNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
AU-rich element binding
protein kinase binding
mRNA 3'-UTR AU-rich region binding
protein homodimerization activity
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19015432.hg.1 | ENST00000351593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015432.hg.1 | ENST00000407627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015432.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015432.hg.1 | TC19001125.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000595499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000595499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000595499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000595499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000595499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000595499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011854.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | TC1900011854.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203841.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | NM_001419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | TC1900011854.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000407627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000593807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000596154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | ENST00000596459.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203842.hg.1 | NM_001419.2 | Refseq |

ELAVL1
Genomic Map