| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001470.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001470.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001470.hg.1 |
TC01000092.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001468.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001468.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001468.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001468.hg.1 |
TC01000092.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001469.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001469.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001469.hg.1 |
TC01000092.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001467.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001467.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001467.hg.1 |
TC01000092.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146386.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146386.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146386.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146386.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146386.hg.1 |
TC0100006698.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100146389.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146389.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146389.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146389.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146389.hg.1 |
TC0100006698.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100146391.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146391.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146391.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146391.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146391.hg.1 |
TC0100006698.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146388.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146388.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146388.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146388.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146388.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146388.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146388.hg.1 |
TC0100006698.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100146387.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146387.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146387.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146387.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146387.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146387.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146387.hg.1 |
TC0100006698.hg.1 |
Clariom D TC |