Assay Details
| Gene Symbol: GUCY2EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024042.1 | 2 - 3 | 453 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 32 | LRRC32 |
D11S833E, GARP |
-47.0kb | Sense | Chr. 11 - 76657441 - 76670941 on NCBI Build GRCh38 |
| Downstream |
tsukushi, small leucine rich proteoglycan |
TSKU |
E2IG4, LRRC54, TSK |
62.4kb | Anti-sense | Chr. 11 - 76782313 - 76798154 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002117.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025291.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025291.hg.1 | TC11002117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025294.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025294.hg.1 | TC11002117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025290.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025290.hg.1 | TC11002117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025292.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025292.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025292.hg.1 | TC11002117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025293.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025293.hg.1 | TC11002117.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000526984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | ENST00000531117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011695.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190263.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190263.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190263.hg.1 | TC1100011695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100190264.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100190264.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190264.hg.1 | ENST00000525328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100190264.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190264.hg.1 | TC1100011695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100190262.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190262.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190262.hg.1 | TC1100011695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100190266.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190266.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190266.hg.1 | TC1100011695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100190265.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190265.hg.1 | NR_024042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100190265.hg.1 | TC1100011695.hg.1 | Clariom D TC |

GUCY2EP
Genomic Map