| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016353.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016353.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016353.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016353.hg.1 |
TC14001454.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016354.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016354.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016354.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016354.hg.1 |
TC14001454.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010019.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174115.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174115.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174115.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174115.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174115.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174115.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174115.hg.1 |
TC1400010019.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400174114.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174114.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174114.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174114.hg.1 |
ENST00000298894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174114.hg.1 |
ENST00000556883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174114.hg.1 |
NM_022151.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174114.hg.1 |
TC1400010019.hg.1 |
Clariom D TC |