Assay Details
| Gene Symbol: NAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128931.1 | NP_001122403.1 | 5 - 6 | 1076 | 99 | |
| NM_138386.2 | NP_612395.2 | 5 - 6 | 1076 | 99 | ||
| XM_011532410.2 | XP_011530712.1 | 5 - 6 | 4543 | 99 | ||
| XM_011532411.2 | XP_011530713.1 | 6 - 7 | 1150 | 99 | ||
| GenBank mRNA | AK097352.1 | - | 5 - 6 | 986 | 99 | |
| AK308601.1 | - | 5 - 6 | 1045 | 99 | ||
| BC008207.1 | - | 5 - 6 | 1076 | 3447276 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001697.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04023461.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023461.hg.1 | ENST00000422287.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023461.hg.1 | ENST00000509434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023461.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023461.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023461.hg.1 | TC04001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191286.hg.1 | TC0400012987.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |

NAF1
Genomic Map