Assay Details
| Gene Symbol: NAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138386.2 | NP_612395.2 | 7 - 8 | 1223 | 124 | |
| XM_011532410.2 | XP_011530712.1 | 7 - 8 | 4690 | 124 | ||
| XM_011532411.2 | XP_011530713.1 | 8 - 9 | 1297 | 124 | ||
| GenBank mRNA | BC008207.1 | - | 7 - 8 | 1223 | 3447276 | 124 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001697.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023454.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023454.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023454.hg.1 | TC04001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023456.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023456.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023456.hg.1 | TC04001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023457.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023457.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023457.hg.1 | TC04001697.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400191281.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191281.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191281.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191281.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191281.hg.1 | TC0400012987.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191280.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191280.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191280.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191280.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191280.hg.1 | TC0400012987.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000422287.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000502973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_001128931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012987.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191282.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191282.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191282.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191282.hg.1 | ENST00000274054.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191282.hg.1 | ENST00000509884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191282.hg.1 | NM_138386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191282.hg.1 | TC0400012987.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 159012 | - | - |

NAF1
Genomic Map