| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008841.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008841.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008841.hg.1 |
TC10000751.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008839.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008839.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008839.hg.1 |
TC10000751.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008840.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008840.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008840.hg.1 |
TC10000751.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000159571.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159571.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159571.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159571.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159571.hg.1 |
TC1000008715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000159567.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159567.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159567.hg.1 |
TC1000008715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000159570.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159570.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159570.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159570.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159570.hg.1 |
TC1000008715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000159568.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159568.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159568.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159568.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159568.hg.1 |
TC1000008715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000159569.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159569.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159569.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159569.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159569.hg.1 |
TC1000008715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
ENST00000370005.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008715.hg.1 |
NM_152310.2 |
Refseq |