Assay Details
| Gene Symbol: CMTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015050.2 | NP_055865.1 | 16 - 17 | 1939 | 68 | |
| XM_005248955.3 | XP_005249012.1 | 16 - 17 | 1869 | 68 | ||
| GenBank mRNA | AK292564.1 | - | 16 - 17 | 1793 | 68 | |
| BC010731.2 | - | 1 - 2 | 66 | 68 | ||
| BC031890.1 | - | 16 - 17 | 1833 | 4944457 | 68 | |
| D43949.2 | - | 16 - 17 | 1829 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000545.hg.1
Set Membership: |
|

CMTR1
Genomic Map