Assay Details
| Gene Symbol: USP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264823.2 | XP_005264880.1 | 7 - 8 | 1449 | 61 | |
| XM_005264824.2 | XP_005264881.1 | 7 - 8 | 1468 | 61 | ||
| XM_005264826.2 | XP_005264883.1 | 7 - 8 | 1449 | 61 | ||
| XM_005264827.2 | XP_005264884.1 | 7 - 8 | 1404 | 61 | ||
| XM_005264829.2 | XP_005264886.1 | 7 - 8 | 1449 | 61 | ||
| XM_005264830.2 | XP_005264887.1 | 6 - 7 | 1146 | 61 | ||
| XM_006712946.2 | XP_006713009.1 | 7 - 8 | 1446 | 61 | ||
| XM_006712949.2 | XP_006713012.1 | 7 - 8 | 1299 | 61 | ||
| XM_017005618.1 | XP_016861107.1 | 7 - 8 | 1449 | 61 | ||
| XM_017005622.1 | XP_016861111.1 | 7 - 8 | 1404 | 61 | ||
| XM_017005627.1 | XP_016861116.1 | 6 - 7 | 1101 | 61 | ||
| XM_017005630.1 | XP_016861119.1 | 6 - 7 | 1146 | 61 | ||
| XM_017005632.1 | XP_016861121.1 | 6 - 7 | 1146 | 61 | ||
| XM_017005634.1 | XP_016861123.1 | 6 - 7 | 1101 | 61 | ||
More Information
Set Membership: |
|

USP19
Genomic Map