Assay Details
| Gene Symbol: CENPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022909.3 | NP_075060.1 | 1 - 2 | 220 | 138 | |
| GenBank mRNA | AB035124.1 | - | 1 - 2 | 133 | 138 | |
| AK123761.1 | - | 1 - 2 | 195 | 138 | ||
| AK290746.1 | - | 1 - 2 | 203 | 138 | ||
| BC012024.1 | - | 1 - 2 | 196 | 4510607 | 138 | |
| BC015355.1 | - | 1 - 2 | 202 | 3451767 | 138 | |
| KU178758.1 | - | 1 - 2 | 133 | 138 | ||
| KU178759.1 | - | 1 - 2 | 133 | 138 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000299.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05003914.hg.1 | ENST00000283006.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003914.hg.1 | ENST00000515001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003914.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003914.hg.1 | TC05000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003915.hg.1 | ENST00000283006.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003915.hg.1 | ENST00000515001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003915.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003915.hg.1 | TC05000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003917.hg.1 | ENST00000283006.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003917.hg.1 | ENST00000515001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003917.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003917.hg.1 | TC05000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003916.hg.1 | ENST00000283006.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003916.hg.1 | ENST00000515001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003916.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003916.hg.1 | TC05000299.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151907.hg.1 | TC0500007666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151905.hg.1 | TC0500007666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151904.hg.1 | TC0500007666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151902.hg.1 | TC0500007666.hg.1 | Clariom D TC |

CENPH
Genomic Map