Assay Details
| Gene Symbol: MED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270629.1 | NP_001257558.1 | 4 - 5 | 376 | 101 | |
| NM_014166.3 | NP_054885.1 | 4 - 5 | 482 | 101 | ||
| XM_017020549.1 | XP_016876038.1 | 4 - 5 | 1570 | 101 | ||
| GenBank mRNA | AF161475.1 | - | 4 - 5 | 444 | 101 | |
| AF230381.1 | - | 4 - 5 | 419 | 101 | ||
| AF300618.1 | - | 4 - 5 | 419 | 101 | ||
| AK001934.1 | - | 4 - 5 | 445 | 101 | ||
| AK222713.1 | - | 4 - 5 | 445 | 101 | ||
| AK301835.1 | - | 4 - 5 | 350 | 101 | ||
| BC005189.1 | - | 4 - 5 | 456 | 3929797 | 101 | |
| CR457276.1 | - | 4 - 5 | 419 | 101 | ||
| CR541830.1 | - | 4 - 5 | 419 | 101 | ||
| HG518167.1 | - | 4 - 5 | 482 | 101 | ||
More Information
Set Membership: |
|

MED4
Genomic Map