Assay Details
| Gene Symbol: PXDNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144651.4 | NP_653252.3 | 21 - 22 | 4247 | 61 | |
| XM_005251168.3 | XP_005251225.1 | 7 - 8 | 2394 | 61 | ||
| XM_006716420.3 | XP_006716483.1 | 19 - 20 | 5545 | 61 | ||
| XM_011517456.2 | XP_011515758.1 | 20 - 21 | 5761 | 61 | ||
| XM_011517457.2 | XP_011515759.1 | 20 - 21 | 5761 | 61 | ||
| XM_011517458.2 | XP_011515760.1 | 19 - 20 | 3921 | 61 | ||
| XM_017013041.1 | XP_016868530.1 | 18 - 19 | 3882 | 61 | ||
| GenBank mRNA | AK058200.1 | - | 5 - 6 | 1467 | 61 | |
| AY877349.1 | - | 21 - 22 | 4247 | 61 | ||
| DB337228.1 | - | 2 - 3 | 61 | |||
| EU170240.1 | - | 21 - 22 | 4146 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001204.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015459.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015459.hg.1 | ENST00000543296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015459.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015459.hg.1 | TC08001204.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800170655.hg.1 | TC0800010410.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000356297.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000519183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522628.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | ENST00000522933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010410.hg.1 | NM_144651.4 | Refseq |

PXDNL
Genomic Map