Assay Details
| Gene Symbol: XRCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318012.1 | NP_001304941.1 | 6 - 7 | 1012 | 132 | |
| NM_001318013.1 | NP_001304942.1 | 6 - 7 | 965 | 132 | ||
| NM_003401.4 | NP_003392.1 | 6 - 7 | 965 | 132 | ||
| NM_022406.3 | NP_071801.1 | 6 - 7 | 965 | 132 | ||
| NM_022550.3 | NP_072044.1 | 6 - 7 | 1012 | 132 | ||
| XM_011543626.1 | XP_011541928.1 | 6 - 7 | 1364 | 132 | ||
| XM_017009827.1 | XP_016865316.1 | 6 - 7 | 965 | 132 | ||
| XM_017009828.1 | XP_016865317.1 | 6 - 7 | 965 | 132 | ||
| GenBank mRNA | AB017445.1 | - | 6 - 7 | 789 | 132 | |
| AK290739.1 | - | 6 - 7 | 793 | 132 | ||
| BC005259.1 | - | 6 - 7 | 817 | 3838919 | 132 | |
| BC016314.1 | - | 6 - 7 | 832 | 4108822 | 132 | |
| BT007216.1 | - | 5 - 6 | 748 | 132 | ||
| U40622.1 | - | 6 - 7 | 798 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000411.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
double-strand break repair via nonhomologous end joining
DNA recombination
response to X-ray
protein sumoylation
DNA ligation involved in DNA repair
positive regulation of ligase activity
cellular response to lithium ion
establishment of integrated proviral latency
DNA binding
protein binding
protein C-terminus binding
ligase activity
double-strand break repair
double-strand break repair via nonhomologous end joining
DNA recombination
response to X-ray
protein sumoylation
DNA ligation involved in DNA repair
positive regulation of ligase activity
cellular response to lithium ion
establishment of integrated proviral latency
DNA binding
protein binding
protein C-terminus binding
ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | ENST00000282268.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | ENST00000338635.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | ENST00000396027.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | ENST00000514538.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005920.hg.1 | TC05000411.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500154574.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154574.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154574.hg.1 | TC0500007966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154576.hg.1 | TC0500007966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000282268.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000338635.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000396027.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000509268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000511817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | ENST00000542685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_003401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022406.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007966.hg.1 | NM_022550.3 | Refseq |

XRCC4
Genomic Map