| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000271357.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367835.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000367838.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000478868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000478868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000478868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000478868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000485232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000485232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000485232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000493800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000493800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000493800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000493800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000493800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000537209.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000539777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
ENST00000546300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267610.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267611.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267612.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267613.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_001267614.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016322.hg.1 |
NM_153832.2 |
Refseq |