| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000269392.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000374782.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000450824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000575907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
NM_001009811.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
NM_014984.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
TC1700012025.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000269392.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000374782.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000450824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
ENST00000575907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
NM_001009811.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198123.hg.1 |
NM_014984.3 |
Refseq |