Assay Details
| Gene Symbol: FAM65C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290268.1 | NP_001277197.1 | 4 - 5 | 752 | 67 | |
| NM_080829.3 | NP_543019.2 | 4 - 5 | 685 | 67 | ||
| NR_110890.1 | 4 - 5 | 937 | 67 | |||
| XM_005260294.3 | XP_005260351.1 | 5 - 6 | 749 | 67 | ||
| XM_006723713.3 | XP_006723776.1 | 4 - 5 | 797 | 67 | ||
| XM_011528578.2 | XP_011526880.1 | 5 - 6 | 777 | 67 | ||
| XM_011528579.2 | XP_011526881.1 | 5 - 6 | 915 | 67 | ||
| XM_011528580.2 | XP_011526882.1 | 5 - 6 | 969 | 67 | ||
| XM_011528581.2 | XP_011526883.1 | 5 - 6 | 953 | 67 | ||
| XM_011528584.2 | XP_011526886.1 | 4 - 5 | 814 | 67 | ||
| XM_011528586.2 | XP_011526888.1 | 4 - 5 | 815 | 67 | ||
| XM_017027682.1 | XP_016883171.1 | 4 - 5 | 814 | 67 | ||
| GenBank mRNA | AK056792.1 | - | 4 - 5 | 656 | 67 | |
| AK295781.1 | - | 4 - 5 | 628 | 67 | ||
| AK299337.1 | - | 4 - 5 | 601 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

FAM65C
Genomic Map