| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
ENST00000045083.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
ENST00000327979.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
ENST00000462493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
NM_001290268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
NM_080829.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
NR_110890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
TC2000009433.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
ENST00000045083.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
ENST00000327979.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
ENST00000462493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
NM_001290268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
NM_080829.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000165997.hg.1 |
NR_110890.1 |
Refseq |