Assay Details
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016569.3 | NP_057653.3 | 3 - 4 | 1673 | 80 | |
| GenBank mRNA | AF216750.1 | - | 3 - 4 | 825 | 80 | |
| BC025258.1 | - | 3 - 4 | 1099 | 5458526 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002000.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
blood vessel development
in utero embryonic development
heart looping
outflow tract morphogenesis
atrioventricular bundle cell differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell aging
positive regulation of cell proliferation
anterior/posterior axis specification, embryo
organ morphogenesis
specification of organ position
stem cell population maintenance
limbic system development
male genitalia development
female genitalia development
negative regulation of epithelial cell differentiation
mammary gland development
luteinizing hormone secretion
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
forelimb morphogenesis
embryonic digit morphogenesis
negative regulation of apoptotic process
negative regulation of myoblast differentiation
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
follicle-stimulating hormone secretion
mesoderm morphogenesis
palate development
ventricular septum morphogenesis
branching involved in mammary gland duct morphogenesis
mammary placode formation
cardiac muscle cell fate commitment
sinoatrial node cell development
cellular senescence
positive regulation of stem cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
protein binding
sequence-specific DNA binding
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
blood vessel development
in utero embryonic development
heart looping
outflow tract morphogenesis
atrioventricular bundle cell differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell aging
positive regulation of cell proliferation
anterior/posterior axis specification, embryo
organ morphogenesis
specification of organ position
stem cell population maintenance
limbic system development
male genitalia development
female genitalia development
negative regulation of epithelial cell differentiation
mammary gland development
luteinizing hormone secretion
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
forelimb morphogenesis
embryonic digit morphogenesis
negative regulation of apoptotic process
negative regulation of myoblast differentiation
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
follicle-stimulating hormone secretion
mesoderm morphogenesis
palate development
ventricular septum morphogenesis
branching involved in mammary gland duct morphogenesis
mammary placode formation
cardiac muscle cell fate commitment
sinoatrial node cell development
cellular senescence
positive regulation of stem cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
protein binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12026251.hg.1 | ENST00000257566.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026251.hg.1 | ENST00000349155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026251.hg.1 | ENST00000361100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026251.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026251.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026251.hg.1 | TC12002000.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000552054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012016.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000257566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000349155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000548503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | ENST00000613550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | NM_005996.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | NM_016569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193645.hg.1 | TC1200012016.hg.1 | Clariom D TC |

TBX3
Genomic Map