Assay Details
| Gene Symbol: UCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003356.3 | NP_003347.1 | 6 - 7 | 1174 | 93 | |
| GenBank mRNA | AF001787.1 | - | 6 - 7 | 1002 | 93 | |
| AF011449.1 | - | 5 - 6 | 917 | 93 | ||
| BC008392.1 | - | 4 - 5 | 713 | 4291158 | 93 | |
| KU178371.1 | - | 3 - 4 | 510 | 93 | ||
| KU178372.1 | - | 5 - 6 | 819 | 93 | ||
| U84763.1 | - | 6 - 7 | 972 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002075.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to superoxide
response to hypoxia
translation
lipid metabolic process
fatty acid metabolic process
mitochondrial transport
aging
response to nutrient
respiratory gaseous exchange
response to cold
response to activity
proton transport
response to insulin
cellular response to hormone stimulus
response to glucocorticoid
structural constituent of ribosome
transporter activity
protein binding
oxidative phosphorylation uncoupler activity
response to superoxide
response to hypoxia
translation
lipid metabolic process
fatty acid metabolic process
mitochondrial transport
aging
response to nutrient
respiratory gaseous exchange
response to cold
response to activity
proton transport
response to insulin
cellular response to hormone stimulus
response to glucocorticoid
structural constituent of ribosome
transporter activity
protein binding
oxidative phosphorylation uncoupler activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000545271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | ENST00000545271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024858.hg.1 | ENST00000314032.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024858.hg.1 | ENST00000348534.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024858.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024858.hg.1 | TC11002075.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024859.hg.1 | ENST00000314032.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024859.hg.1 | ENST00000348534.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024859.hg.1 | ENST00000545271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024859.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024859.hg.1 | TC11002075.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000426995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000544614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000545271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | ENST00000545271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011605.hg.1 | NM_022803.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189498.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189498.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189498.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189498.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189498.hg.1 | TC1100011605.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100189499.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189499.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189499.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189499.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189499.hg.1 | TC1100011605.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100189500.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189500.hg.1 | ENST00000545271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189500.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189500.hg.1 | ENST00000314032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189500.hg.1 | ENST00000545271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189500.hg.1 | NM_003356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189500.hg.1 | TC1100011605.hg.1 | Clariom D TC |

UCP3
Genomic Map