| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05009225.hg.1 |
ENST00000297156.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05009225.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05009225.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05009225.hg.1 |
TC05000667.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500159765.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159765.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159765.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500159765.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159765.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159765.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500159765.hg.1 |
TC0500008702.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000297156.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
ENST00000514518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008702.hg.1 |
NM_001745.3 |
Refseq |