Assay Details
| Gene Symbol: PCDHGA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018918.2 | NP_061741.1 | 1 - 2 | 2423 | 140 | |
| GenBank mRNA | AF152325.1 | - | 1 - 2 | 2423 | 140 | |
More Information
Set Membership: |
|

PCDHGA5
Genomic Map