Assay Details
| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021137.4 | NP_066960.1 | 1 - 2 | 334 | 91 | |
| XM_017024993.1 | XP_016880482.1 | 1 - 2 | 290 | 91 | ||
| GenBank mRNA | AK312387.1 | - | 1 - 2 | 95 | 91 | |
| BC001643.2 | - | 1 - 2 | 81 | 2823154 | 91 | |
| BC001949.1 | - | 1 - 2 | 81 | 2823154 | 91 | |
| BC003694.2 | - | 1 - 2 | 96 | 91 | ||
| DB093553.1 | - | 1 - 2 | 334 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000320.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312387.1,BC001643.2,BC001949.1,NM_021137.4,XM_017024993.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
apoptotic process
immune response
embryo development
cell migration
protein ubiquitination
negative regulation of Rho protein signal transduction
stress fiber assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA replication
protein homooligomerization
ubiquitin-protein transferase activity
protein binding
GTP-Rho binding
protein domain specific binding
DNA replication
apoptotic process
immune response
embryo development
cell migration
protein ubiquitination
negative regulation of Rho protein signal transduction
stress fiber assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA replication
protein homooligomerization
ubiquitin-protein transferase activity
protein binding
GTP-Rho binding
protein domain specific binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003844.hg.1 | ENST00000226225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003844.hg.1 | ENST00000544907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003844.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003844.hg.1 | TC17000320.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003845.hg.1 | ENST00000226225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003845.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003845.hg.1 | TC17000320.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003846.hg.1 | ENST00000226225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003846.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003846.hg.1 | TC17000320.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003841.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003841.hg.1 | TC17000320.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003842.hg.1 | ENST00000226225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003842.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003842.hg.1 | TC17000320.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700152293.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152293.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152293.hg.1 | TC1700007361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152293.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152293.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | TC1700007361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152295.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | TC1700007361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152298.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | TC1700007361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152301.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | TC1700007361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152300.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152292.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152292.hg.1 | TC1700007361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152292.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152294.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152294.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152294.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152294.hg.1 | TC1700007361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152294.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152294.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152294.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |

TNFAIP1
Genomic Map