| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019770.hg.1 |
ENST00000381007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019770.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019770.hg.1 |
TC05001411.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019769.hg.1 |
ENST00000381007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019769.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019769.hg.1 |
TC05001411.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500176274.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176274.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176274.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500176274.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176274.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176274.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500176274.hg.1 |
TC0500010932.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
ENST00000506816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010932.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500176276.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176276.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500176276.hg.1 |
ENST00000381007.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176276.hg.1 |
NM_019072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500176276.hg.1 |
TC0500010932.hg.1 |
Clariom D TC |