Assay Details
| Gene Symbol: EEF2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013302.3 | NP_037434.1 | 13 - 14 | 1923 | 82 | |
| XM_017023193.1 | XP_016878682.1 | 13 - 14 | 1634 | 82 | ||
| GenBank mRNA | BC032665.1 | - | 14 - 15 | 1634 | 5553055 | 82 |
| U93850.1 | - | 12 - 13 | 1438 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000243.hg.1
Set Membership: |
|

EEF2K
Genomic Map